Would you like to log in again?
DNase Test Agar with Toluidine Blue, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
CRITERION™
C5670 C5672 C5670 C5671 C5673 C5673 C5671 C5672
89405-404EA 128.76 USD InStock
89405-404 CA89405-408 CA89405-404 CA89405-406 89405-410 CA89405-410 89405-406 89405-408
DNase Test Agar with Toluidine Blue, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ DNase Test Agar with Toluidine Blue is recommended for the detection of DNase in gram-negative bacteria, especially Serratiaspp
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty