Water for molecular biology, DEPC treated

Synonyms: Dihydrogen oxide

CG480BX
CA89407-022
Water for molecular biology, DEPC treated
Water

CulGenex™ DEPC Treated, Molecular Grade Water is chemically treated for use in various applications in molecular biology, particularly for creating RNAse-free solutions or to rinse RNAses from labware.

Formula: H₂O
MW: 18.02 g/mol
MDL Number: MFCD00011332
CAS Number: 7732-18-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR