    Sort by:
  • Search Within Results
D(+)-Talose
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"1"
Sort by:
Supplier:  TCI America
Description:   CAS Number: 2595-98-4 MDL Number: MFCD00135834 Molecular Formula: C6H12O6 Molecular Weight: 180.16 Purity/Analysis Method: 98.0% (HPLC) Form:...