PuriCare® Procedure Stations, Domestic Electrical Configurations, Labconco®
31141010 31141100 31241000 31141000 31261010 31141110 31261100 31241010 31161110 31241110 31261110 31161010 31261000 31241100 31161000 31161100
89414-438EA 13479.91 USD
89414-438 89414-404 89414-402 89414-400 89414-488 89414-444 89414-486 89414-398 89414-442 89414-396 89414-440 89414-492 89414-394 89414-490 89414-392 89414-390
PuriCare® Procedure Stations, Domestic Electrical Configurations, Labconco®
PuriCare Procedure Stations provide personnel, animal and environmental protection
They are specialized Class II A1 or A2 biological safety cabinets that meet or exceed the product and personnel protection requirements of NSF 49. These stations are designed for conducting small animal research activities including cage changing, weighing, anesthesia delivery and surgical procedures. Please refer to sash opening, electrical configuration and product specifications for details.
Certifications: Models with 10" sash openings for 115 volt operation are NSF Listed