Purifier® Cell Logic®+ Class II B2 Biosafety Cabinets, Domestic Electrical Configurations, Labconco®
Supplier: Labconco
343481010 343681110 323681100 323681010 333481110 333681110 323481110 333681100 343481000 323681110 343481110 323481000 323681000 333481100 343681100 333481000 323481010 333681010 333681000 333481010 323481100 343481100 343681000 343681010
89413-994EA 25260.02 USD InStock
89413-994 89414-008 89413-896 89414-014 89414-000 89414-012 89414-004 89413-892 89413-874 89414-016 89413-996 89413-882 89413-894 89413-880 89413-888 89413-878 89414-002 89414-010 89413-890 89413-998 89413-884 89413-876 89413-886 89414-006
Purifier® Cell Logic®+ Class II B2 Biosafety Cabinets, Domestic Electrical Configurations, Labconco®
Fume Hoods and Enclosures Biological and Microbiological Safety Cabinets
The new Purifier Cell Logic+ Class II B2 Biosafety Cabinet excels in user comfort and functionality
Using the same ECM blower technology as the Purifier Logic®, the Cell Logic+ uses the Constant Airflow Profile™ (CAP) to maintain a precise volume of air as the filters load. In addition, the cabinet will continuously display the HEPA filter life remaining and cabinet status. These features combine to provide unparalleled safety.
Cell Logic+ B2 Biosafety Cabinets, designed to be hard-ducted to the outside, are equipped to meet the specific needs of the cell culture industry. Optional features include the Scope-Ready™ and Temp-Zone™ packages. The Scope-Ready package allows the user’s microscope to be integrated into the biosafety cabinet. The Temp-Zone package includes a temperature controlled area on the work surface. The Cell Logic+ is available with the Scope-Ready package, the Temp-Zone package or both. Some cabinets include base stands and/or additional options. Please refer to sash opening, electrical configuration and product specifications for details
Decades of industry leading advances in ergonomic design have shaped the Inclination™ Technology Suite that includes the following: 10° angled sash, patented curved air grille with built in armrest, fully accessible controls and eye-level mounted color display with MyLogic™ operating system. MyLogic provides intuitive programming, color coded menus, status messages and alarms with diagnostic troubleshooting—all using full sentences.