Purifier® Logic®+ Class II B2 Biosafety Cabinets, Domestic Electrical Configurations, Labconco®
303680010 303480010 303481110 303681000 303681110 303480100 303481100 303681010 303480110 303681100 303680100 303680000 303480000 303481000 303481010 303680110
89412-960EA 23377.44 USD
89412-960 89412-950 89412-920 89412-954 89412-952 89412-946 89412-924 89412-914 89412-958 89412-922 89412-956 89412-928 89412-918 89412-926 89412-948 89412-916
Purifier® Logic®+ Class II B2 Biosafety Cabinets, Domestic Electrical Configurations, Labconco®
The new Purifier Logic+ Class II B2 Biosafety Cabinet excels in user comfort and functionality
Using the same ECM blower technology as the Purifier Logic®, the Logic+ uses the Constant Airflow Profile™ (CAP) to maintain a precise volume of air as the filters load. In addition, the cabinet will continuously display the HEPA filter life remaining and cabinet status. These features combine to provide unparalleled safety.
These biosafety cabinets are designed to be hard ducted to the outside. The cabinet provides personnel, product and environmental protection and is suitable for work in biosafety levels 1, 2 and 3. In addition, some models have base stands, UV lights, service fixtures, pass throughs and/or a combination of those features included. Please refer to sash opening, electrical configuration and product specifications for details
Decades of industry leading advances in ergonomic design have shaped the Inclination™ Technology Suite that includes the following: 10° angled sash, patented curved air grille with built in armrest, fully accessible controls and eye-level mounted color display with MyLogic™ operating system. MyLogic provides intuitive programming, color coded menus, status messages and alarms with diagnostic troubleshooting—all using full sentences.