Purifier® Cell Logic®+ Class II A2 Biosafety Cabinets, International Electrical Configurations, Labconco®
Supplier: Labconco
322591150
89414-172EA 17094.26 USD
89414-172
Purifier® Cell Logic®+ Class II A2 Biosafety Cabinets, International Electrical Configurations, Labconco®
The new Purifier Cell Logic+ Class II A2 Biosafety Cabinet excels in user comfort and functionality
Using the same ECM blower technology as the Purifier Logic®, the Cell Logic+ uses the Constant Airflow Profile™ (CAP) to maintain a precise volume of air as the filters load. In addition, the cabinet will continuously display the HEPA filter life remaining and cabinet status. These features combine to provide unparalleled safety.
Cell Logic+ A2 Biosafety Cabinets, designed to discharge HEPA-filtered exhaust air directly into the laboratory or into an exhaust system through the optional Canopy Connection, are equipped to meet the specific needs of the cell culture industry. Optional features include the Scope-Ready™ and Temp-Zone™ packages. The Scope-Ready package allows the user’s microscope to be integrated into the biosafety cabinet. The Temp-Zone package includes a temperature controlled area on the work surface. The Cell Logic+ is available with the Scope-Ready package, the Temp-Zone package or both. Some cabinets include base stands and/or additional options. Please refer to sash opening, electrical configuration and product specifications for details
Decades of industry leading advances in ergonomic design have shaped the Inclination™ Technology Suite that includes the following: 10° angled sash, patented curved air grille with built in armrest, fully accessible controls and eye-level mounted color display with MyLogic™ operating system. MyLogic provides intuitive programming, color coded menus, status messages and alarms with diagnostic troubleshooting—all using full sentences.