Would you like to log in again?
Purifier® Cell Logic®+ Class II A2 Biosafety Cabinets, Domestic Electrical Configurations, Labconco®
Supplier: Labconco
332391110 322691010 322691110 332691110 332391010 322391010 342691010 342691110 332691010 322591100 322591000 322391110 342391010 342391110 322491100 342591010 332591010 332591110 342591110 342491000 342491100 332491100 322491000 332491000 332391000 332691100 332691000 322691000 322691100 342691100 342691000 322591110 322591010 322391100 322391000 342391000 342391100 332391100 342591100 342591000 332591000 332591100 342491110 342491010 322491010 332491110 322491110 332491010
The new Purifier Cell Logic+ Class II A2 Biosafety Cabinet excels in user comfort and functionality
Using the same ECM blower technology as the Purifier Logic®, the Cell Logic+ uses the Constant Airflow Profile™ (CAP) to maintain a precise volume of air as the filters load. In addition, the cabinet will continuously display the HEPA filter life remaining and cabinet status. These features combine to provide unparalleled safety.
Cell Logic+ A2 Biosafety Cabinets, designed to discharge HEPA-filtered exhaust air directly into the laboratory or into an exhaust system through the optional Canopy Connection, are equipped to meet the specific needs of the cell culture industry. Optional features include the Scope-Ready™ and Temp-Zone™ packages. The Scope-Ready package allows the user’s microscope to be integrated into the biosafety cabinet. The Temp-Zone package includes a temperature controlled area on the work surface. The Cell Logic+ is available with the Scope-Ready package, the Temp-Zone package or both. Some cabinets include base stands and/or additional options. Please refer to sash opening, electrical configuration and product specifications for details
Decades of industry leading advances in ergonomic design have shaped the Inclination™ Technology Suite that includes the following: 10° angled sash, patented curved air grille with built in armrest, fully accessible controls and eye-level mounted color display with MyLogic™ operating system. MyLogic provides intuitive programming, color coded menus, status messages and alarms with diagnostic troubleshooting—all using full sentences.
Services: VWRCATALYST™ offers customer on-site validation, Testing & certification services including Face Velocity Profile Test, Smoke Generation Test, Tracer Gas Test, IQ, OQ, and PQ. Please call 888-793-2300 or email VWRCATALYST™@vwr.com for more details. You can also visit https://vwr.com/equip_services.
ORDER
Expand All / Collapse All
Description Electrical Depth Height Width Includes Supplier No. VWR Catalog Number Unit Price Quantity
3' Purifier Cell Logic+ A2 Biosafety Cabinet with Service Fixture and Pass Through Only 115V 31 3/16" 61 13/16" 42 5/16" Scope Ready Package 332391000 89414-046 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
3' Purifier Cell Logic+ A2 Biosafety Cabinet with Service Fixture and Pass Through Only 115V 31 3/16" 61 13/16" 42 5/16" Scope Ready Package, Temp Zone Package 342391000 89414-042 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
3' Purifier Cell Logic+ A2 Biosafety Cabinet with Service Fixture and Pass Through Only 230V NA 31 3/16" 61 13/16" 42 5/16" Scope Ready Package 332391010 89414-286 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
3' Purifier Cell Logic+ A2 Biosafety Cabinet with Service Fixture and Pass Through Only 230V NA 31 3/16" 61 13/16" 42 5/16" Scope Ready Package, Temp Zone Package 342391010 89414-282 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
3' Purifier Cell Logic+ A2 Biosafety Cabinet with Telescoping Base Stand, Service Fixture, and Pass Through Only 115V 31 3/16" 89 5/16" 42 5/16" Scope Ready Package 332391100 89414-048 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
3' Purifier Cell Logic+ A2 Biosafety Cabinet with Telescoping Base Stand, Service Fixture, and Pass Through Only 115V 31 3/16" 89 5/16" 42 5/16" Scope Ready Package, Temp Zone Package 342391100 89414-044 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
3' Purifier Cell Logic+ A2 Biosafety Cabinet with Telescoping Base Stand, Service Fixture, and Pass Through Only 230V NA 31 3/16" 89 5/16" 42 5/16" Scope Ready Package 332391110 89414-288 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
3' Purifier Cell Logic+ A2 Biosafety Cabinet with Telescoping Base Stand, Service Fixture, and Pass Through Only 230V NA 31 3/16" 89 5/16" 42 5/16" Scope Ready Package, Temp Zone Package 342391110 89414-284 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
3' Purifier Cell Logic+ A2 Biosafety Cabinet with Telescoping Base Stand, UV Light, Service Fixtures, and Pass Through 115V 31 3/16" 89 5/16" 42 5/16" Temp Zone Package 322391100 89414-052 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
3' Purifier Cell Logic+ A2 Biosafety Cabinet with Telescoping Base Stand, UV Light, Service Fixtures, and Pass Through 230V NA 31 3/16" 89 5/16" 42 5/16" Temp Zone Package 322391110 89414-292 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
3' Purifier Cell Logic+ A2 Biosafety Cabinet with UV Light, Service Fixtures, and Pass Through 115V 31 3/16" 61 13/16" 42 5/16" Temp Zone Package 322391000 89414-050 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
3' Purifier Cell Logic+ A2 Biosafety Cabinet with UV Light, Service Fixtures, and Pass Through 230V NA 31 3/16" 61 13/16" 42 5/16" Temp Zone Package 322391010 89414-290 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
4' Purifier Cell Logic+ A2 Biosafety Cabinet with Service Fixture and Pass Through Only 115V 31 3/16" 61 13/16" 54 5/16" Scope Ready Package 332491000 89414-058 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
4' Purifier Cell Logic+ A2 Biosafety Cabinet with Service Fixture and Pass Through Only 115V 31 3/16" 61 13/16" 54 5/16" Scope Ready Package, Temp Zone Package 342491000 89414-054 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
4' Purifier Cell Logic+ A2 Biosafety Cabinet with Service Fixture and Pass Through Only 230V NA 31 3/16" 61 13/16" 54 5/16" Scope Ready Package 332491010 89414-298 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
4' Purifier Cell Logic+ A2 Biosafety Cabinet with Service Fixture and Pass Through Only 230V NA 31 3/16" 61 13/16" 54 5/16" Scope Ready Package, Temp Zone Package 342491010 89414-294 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
4' Purifier Cell Logic+ A2 Biosafety Cabinet with Telescoping Base Stand, Service Fixture, and Pass Through Only 115V 31 3/16" 89 5/16" 54 5/16" Scope Ready Package 332491100 89414-060 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
4' Purifier Cell Logic+ A2 Biosafety Cabinet with Telescoping Base Stand, Service Fixture, and Pass Through Only 115V 31 3/16" 89 5/16" 54 5/16" Scope Ready Package, Temp Zone Package 342491100 89414-056 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
4' Purifier Cell Logic+ A2 Biosafety Cabinet with Telescoping Base Stand, Service Fixture, and Pass Through Only 230V NA 31 3/16" 89 5/16" 54 5/16" Scope Ready Package 332491110 89414-300 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
4' Purifier Cell Logic+ A2 Biosafety Cabinet with Telescoping Base Stand, Service Fixture, and Pass Through Only 230V NA 31 3/16" 89 5/16" 54 5/16" Scope Ready Package, Temp Zone Package 342491110 89414-296 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
4' Purifier Cell Logic+ A2 Biosafety Cabinet with Telescoping Base Stand, UV Light, Service Fixtures, and Pass Through 115V 31 3/16" 89 5/16" 54 5/16" Temp Zone Package 322491100 89414-064 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
4' Purifier Cell Logic+ A2 Biosafety Cabinet with Telescoping Base Stand, UV Light, Service Fixtures, and Pass Through 230V NA 31 3/16" 89 5/16" 54 5/16" Temp Zone Package 322491110 89414-304 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
4' Purifier Cell Logic+ A2 Biosafety Cabinet with UV Light, Service Fixtures, and Pass Through 115V 31 3/16" 61 13/16" 54 5/16" Temp Zone Package 322491000 89414-062 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
4' Purifier Cell Logic+ A2 Biosafety Cabinet with UV Light, Service Fixtures, and Pass Through 230V NA 31 3/16" 61 13/16" 54 5/16" Temp Zone Package 322491010 89414-302 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
5' Purifier Cell Logic+ A2 Biosafety Cabinet with Service Fixture and Pass Through Only 115V 31 3/16" 61 13/16" 66 5/16" Scope Ready Package 332591000 89414-070 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
5' Purifier Cell Logic+ A2 Biosafety Cabinet with Service Fixture and Pass Through Only 115V 31 3/16" 61 13/16" 66 5/16" Scope Ready Package, Temp Zone Package 342591000 89414-066 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
5' Purifier Cell Logic+ A2 Biosafety Cabinet with Service Fixture and Pass Through Only 230V NA 31 3/16" 61 13/16" 66 5/16" Scope Ready Package 332591010 89414-310 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
5' Purifier Cell Logic+ A2 Biosafety Cabinet with Service Fixture and Pass Through Only 230V NA 31 3/16" 61 13/16" 66 5/16" Scope Ready Package, Temp Zone Package 342591010 89414-306 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
5' Purifier Cell Logic+ A2 Biosafety Cabinet with Telescoping Base Stand, Service Fixture, and Pass Through Only 115V 31 3/16" 89 5/16" 66 5/16" Scope Ready Package 332591100 89414-072 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
5' Purifier Cell Logic+ A2 Biosafety Cabinet with Telescoping Base Stand, Service Fixture, and Pass Through Only 115V 31 3/16" 89 5/16" 66 5/16" Scope Ready Package, Temp Zone Package 342591100 89414-068 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
5' Purifier Cell Logic+ A2 Biosafety Cabinet with Telescoping Base Stand, Service Fixture, and Pass Through Only 230V NA 31 3/16" 89 5/16" 66 5/16" Scope Ready Package 332591110 89414-312 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
5' Purifier Cell Logic+ A2 Biosafety Cabinet with Telescoping Base Stand, Service Fixture, and Pass Through Only 230V NA 31 3/16" 89 5/16" 66 5/16" Scope Ready Package, Temp Zone Package 342591110 89414-308 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
5' Purifier Cell Logic+ A2 Biosafety Cabinet with Telescoping Base Stand, UV Light, Service Fixtures, and Pass Through 115V 31 3/16" 89 5/16" 66 5/16" Temp Zone Package 322591100 89414-076 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
5' Purifier Cell Logic+ A2 Biosafety Cabinet with Telescoping Base Stand, UV Light, Service Fixtures, and Pass Through 230V NA 31 3/16" 89 5/16" 66 5/16" Temp Zone Package 322591110 89414-316 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
5' Purifier Cell Logic+ A2 Biosafety Cabinet with UV Light, Service Fixtures, and Pass Through 115V 31 3/16" 61 13/16" 66 5/16" Temp Zone Package 322591000 89414-074 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
5' Purifier Cell Logic+ A2 Biosafety Cabinet wih UV Light, Service Fixtures, and Pass Through 230V NA 31 3/16" 61 13/16" 66 5/16" Temp Zone Package 322591010 89414-314 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
6' Purifier Cell Logic+ A2 Biosafety Cabinet with Service Fixture and Pass Through Only 115V 31 3/16" 61 13/16" 66 5/16" Scope Ready Package 332691000 89414-082 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
6' Purifier Cell Logic+ A2 Biosafety Cabinet with Service Fixture and Pass Through Only 115V 31 3/16" 61 13/16" 66 5/16" Scope Ready Package, Temp Zone Package 342691000 89414-078 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
6' Purifier Cell Logic+ A2 Biosafety Cabinet with Service Fixture and Pass Through Only 230V NA 31 3/16" 61 13/16" 66 5/16" Scope Ready Package 332691010 89414-322 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
6' Purifier Cell Logic+ A2 Biosafety Cabinet with Service Fixture and Pass Through Only 230V NA 31 3/16" 61 13/16" 66 5/16" Scope Ready Package, Temp Zone Package 342691010 89414-318 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
6' Purifier Cell Logic+ A2 Biosafety Cabinet with Telescoping Base Stand, Service Fixture, and Pass Through Only 115V 31 3/16" 89 5/16" 66 5/16" Scope Ready Package 332691100 89414-084 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
6' Purifier Cell Logic+ A2 Biosafety Cabinet with Telescoping Base Stand, Service Fixture, and Pass Through Only 115V 31 3/16" 89 5/16" 66 5/16" Scope Ready Package, Temp Zone Package 342691100 89414-080 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
6' Purifier Cell Logic+ A2 Biosafety Cabinet with Telescoping Base Stand, Service Fixture, and Pass Through Only 230V NA 31 3/16" 89 5/16" 66 5/16" Scope Ready Package 332691110 89414-324 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
6' Purifier Cell Logic+ A2 Biosafety Cabinet with Telescoping Base Stand, Service Fixture, and Pass Through Only 230V NA 31 3/16" 89 5/16" 66 5/16" Scope Ready Package, Temp Zone Package 342691110 89414-320 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
6' Purifier Cell Logic+ A2 Biosafety Cabinet with Telescoping Base Stand, UV Light, Service Fixtures, and Pass Through 115V 31 3/16" 89 5/16" 66 5/16" Temp Zone Package 322691100 89414-088 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
6' Purifier Cell Logic+ A2 Biosafety Cabinet with Telescoping Base Stand, UV Light, Service Fixtures, and Pass Through 230V NA 31 3/16" 89 5/16" 66 5/16" Temp Zone Package 322691110 89414-328 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
6' Purifier Cell Logic+ A2 Biosafety Cabinet with UV Light, Service Fixtures, and Pass Through 115V 31 3/16" 61 13/16" 66 5/16" Temp Zone Package 322691000 89414-086 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
6' Purifier Cell Logic+ A2 Biosafety Cabinet with UV Light, Service Fixtures, and Pass Through 230V NA 31 3/16" 61 13/16" 66 5/16" Temp Zone Package 322691010 89414-326 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
SPECIFICATIONS
Environmentally Preferable Attribute Product and/or packaging contains recycled/renewable content, Product is energy and/or water efficient
Sash Opening 9"
SERVICES
Services: VWRCATALYST™ offers customer on-site validation, Testing & certification services including Face Velocity Profile Test, Smoke Generation Test, Tracer Gas Test, IQ, OQ, and PQ. Please call 888-793-2300 or email VWRCATALYST™@vwr.com for more details. You can also visit https://vwr.com/equip_services.
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty