Purifier® Logic®+ Class II A2 Biosafety Cabinets, Domestic Electrical Configurations, Labconco®
302680010 302681110 302411010 302611110 302380110 302680000 302481110 302410110 302480100 302310000 302381110 302311110 302410100 302410010 302380010 302681010 302311010 302511010 302580100 302480000 302610010 302380000 302511110 302310010 302310100 302310110 302510010 302610110 302680110 302611010 302580000 302381010 302581110 302510100 302411110 302610100 302580010 302510000 302580110 302680100 302581010 302410000 302610000 302380100 302481010 302480110 302480010 302510110
89412-582EA 13617.7 USD
89412-582 89412-560 89412-580 89412-564 89412-586 89412-542 89412-540 89412-562 89412-584 89412-546 89412-568 89412-524 89412-602 89412-522 89412-566 89412-544 89412-588 89412-600 89412-616 89412-614 89412-538 89412-590 89412-594 89412-550 89412-572 89412-592 89412-570 89412-576 89412-532 89412-554 89412-598 89412-596 89412-530 89412-552 89412-574 89412-612 89412-558 89412-536 89412-610 89412-578 89412-556 89412-534 89412-528 89412-606 89412-526 89412-548 89412-604 89412-608
Purifier® Logic®+ Class II A2 Biosafety Cabinets, Domestic Electrical Configurations, Labconco®
The cabinet provides personnel, product and environmental protection and is suitable for work in biosafety levels 1, 2 and 3
Using the same ECM blower technology as the Purifier Logic®, the Logic+ uses the Constant Airflow Profile™ (CAP) to maintain a precise volume of air as the filters load. In addition, the cabinet will continuously display the HEPA filter life remaining and cabinet status. These features combine to provide unparalleled safety.
The cabinet provides personnel, product and environmental protection and is suitable for work in biosafety levels 1, 2 and 3. These biohazard cabinets are designed to discharge HEPA-filtered exhaust air directly into the laboratory, or into an exhaust system through the optional Canopy Connection. In addition, some models have base stands, UV lights, service fixtures, pass throughs and/or a combination of those features included. Please refer to sash opening, electrical configuration and product specifications for details
These biohazard cabinets are designed to discharge HEPA-filtered exhaust air directly into the laboratory, or into an exhaust system through the optional Canopy Connection. In addition, some models have base stands, UV lights, service fixtures, pass throughs and/or a combination of those features included. Please refer to sash opening, electrical configuration and product specifications for details.
Accessories information: Ventus Canopy Connection for Purifier Logic and Logic+ Biosafety Cabinets. For thimble ducting Class II, Type A2 Biosafety Cabinets. Canopy provides an air gap to allow room air to enter the ductwork and mix with the filtered air being exhausted to the outside. Inadequate exhaust activates the switch in less than one second and signals the LCD on the cabinet to display an alarm and the motor to increase to maximum speed for personal safety. Adds only 1.25" to the overall height of the cabinet. Powder-coated steel. Outlet is 10" ID. Exhaust connection is required. Damper 82001-060 and Remote Blower 82006-740 or 82006-742 are recommended (not included). Logic models require conversion cable and controller. Note if for Logic or Logic+ when ordering.