Heating Tapes, Ribbon-Type Elements, Ace Glass Incorporated
Supplier: Ace Glass
12064-08 12064-44 12064-06 12064-23 12064-29 12064-42 12064-27 12064-31 12064-25 12064-02 12064-04
89076-208EA 248.41 USD
89076-208 89076-218 89076-206 89076-216 89076-204 89076-214 89076-202 89076-212 89076-200 89076-210 89076-220
Heating Tapes, Ribbon-Type Elements, Ace Glass Incorporated
Tapes
Glas-Col heating tapes use two ribbon-type heating elements running lengthwise in the tape
These heating elements are encased in durable glass braid, then impregnated and covered with tough silicone rubber. This construction method easily passes a 1000 volt test for electrical insulation. Silicone rubber is also used to insulate the stranded terminal leads at one end for waterproof fittings at both ends of the tape. The only limitation is the upper temperature limit of 249 °C (480 °F) for the silicone covering.