Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • GSA Contract (32) MWBE (8) SBE (15)
  • Supplier
  • Suppliers found in search results
    Sort by:

  • Search Within Results

dntps

https://us.vwr.com/stibo/smallweb/std.lang.all/53/60/18945360.jpg

Deoxyribonucleotides are the foundation of DNA, and it takes thousands of these nucleotides to make up strands. DNA consists of sugar molecules called deoxyribose, a nitrogenous base of purine in adenine and guanine or pyrimidine in thymine or cytosine, and a phosphate molecule. dNTP solution mixes are used for high-purity reaction procedures. The mixtures are available in a range of concentrations, specifications, and kit sizes.


SearchPresentationType-VERTICAL
 
 
SearchResultCount:"47"
Sort by:
 
 
 
 

Supplier:  VWR
Description:   Deoxyribonucleotides are the foundation of DNA and it takes thousands of these nucleotides to make up DNA.
SDS
Catalog Number: (97062-862)
Supplier:  VWR
Description:   DATP SOL BIOTECH GRADE 100MM 40UMOLE
SDS
Catalog Number: (97062-864)
Supplier:  VWR
Description:   DCTP SOL BIOTECH GRADE 100MM 40UMOLE
SDS
Catalog Number: (97062-872)
Supplier:  VWR
Description:   DTTP SOL BIOTECH GRADE 100MM 40UMOLE
SDS
Catalog Number: (97062-866)
Supplier:  VWR
Description:   DGTP SOL BIOTECH GRADE 100MM 40UMOLE
SDS
Supplier:  Quantabio
Description:   The 10mM dNTP Mix is a solution of high purity deoxyribonucleoside triphosphates that has been functionally qualified for real-time quantitative PCR (...
SDS
Supplier:  GE Healthcare - Life Sciences
Description:   Easy to use, time saving solutions for in vitro transcription or for reactions requiring rNTPs of high purity.
SDS
Product available on GSA Advantage®
Supplier:  GE Healthcare - Life Sciences
Description:   Greater than 99% triphosphate purity (by HPLC) for optimum performance and consistency
SDS
Product available on GSA Advantage®
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 8  of 47
  1  2  3  4  5  6  Next