    Sort by:
(2-Butenyl)triphenylphosphonium chloride
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Alfa Aesar
Description:   (3-Methylallyl)triphenylphosphonium chloride. Grade: 97, Melting Point ca 225-230*[degree]C. Boiling Point C: NA. C22H22ClP. 13138-25-5. IRRITANT HYGROSCOPIC