    Sort by:
  • Search Within Results
2,2,3,4,4,4-Hexafluorobutyl methacrylate
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"1"
Sort by:
Supplier:  Alfa Aesar
Description:   Methacrylic acid 2,2,3,4,4,4-hexafluorobutyl ester. Grade:97, Melting Point C. Boiling Point C:70*/50mm. C8H8F6O2. 36405-47-7. Stabilised with 50 ppm 4-methoxyphenol. IRRITANT KEEP COLD