    Sort by:
  • Search Within Results
3-Chloropropyl chloroformate
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"1"
Sort by:
Supplier:  TCI America
Description:   CAS Number: 628-11-5 Molecular Formula: C4H6Cl2O2 Molecular Weight: 156.99 Purity/Analysis Method: 95.0% (GC) Form: Clear Liquid Boiling point...