    Sort by:
  • Search Within Results
You Searched for: "4-tert-butylbenzophenone"
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"1"
Sort by:
Supplier:  Alfa Aesar
Description:   . Grade:98, Melting Point C38-40*. Boiling Point C:168-170*/2mm. C17H18O. 22679-54-5.