    Sort by:
  • Search Within Results
You Searched for: "4-benzyloxy-benzaldehyde-on-wang-resin"
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"1"
Sort by:
Catalog Number: (AAAL19370-06)
Supplier:  Alfa Aesar
Description:   Aldehyde Wang resin Benzaldehyde Wang resin. Grade: 0. 8-1. 0 mmol/g on Merrifield resin. Melting Point C. Boiling Point C: NA. 151896-98-9. 2% crosslinked, 200-400 mesh. AIR SENSITIVE STORE UNDER NITROGEN