Multi-Parameter ProLab 4000 Meter, SI Analytics
285203870 285204260
97042-272EA 3713.54 USD InStock
97042-272 97042-348
Multi-Parameter ProLab 4000 Meter, SI Analytics
Electrochemical Meters pH and Multi-Parameter Meters
  • 3 Channel Instrument
  • 2 channels for pH/mV/ISE
  • 1 channel for Conductivity
  • German engineered/manufactured
  • Three Year Warranty
  • Advanced algorithym for faster more precise measurements
  • RS-232/USB
  • Password protection
  • GLP Compliant
The ProLab 4000 precision multi parameter meter enables you to perform pH, ORP, conductivity and ion-selective measurements rapidly and reliably with the ability to measure three parameters simultaneously plus temperature. This instrument is engineered/manufactured in Germany and backed by 65+ years of measuring experience. It provides the maximum degree of ease of use, reliability and, above all, measuring certainty for all applications. Fast, convenient and precise, the instrument can be operated just like a PC via the menu keys or by using a mouse (included). Easy to read, high resolution, backlit, color display.
The ProLab 4000 offers automatic user identification with electronic ID transponder and password as well as wireless sensor recognition. The CalClock provides a unique combination of sensor evaluation and calibration timer control in one easy to read symbol.Perfectly suitable for all GLP protocol. Check out all of our ProLab models to find the one that best fits your lab.
Certifications: CE Certified, UL STD 61010-1 Certified, cETLus Certified.
Ordering information: Meter and Universal Power Supply only. Kit is also available (97042-272). Kit includes ProLab 4000 measuring instrument, mouse, IoLine 3in1 pharmaceutical grade double junction pH electrode (97042-386), wide-range conductivity sensor (97042-222), buffer solutions, electrode arm, universal power supply and cover.