Would you like to log in again?
Multi-Parameter ProLab 3000 Meter, SI Analytics
285203860 285204250
97042-270EA 2962.57 USD InStock
97042-270 97042-346
Multi-Parameter ProLab 3000 Meter, SI Analytics
Electrochemical Meters pH and Multi-Parameter Meters
  • 2 Channels for pH/mV/ISE
  • German engineered/Manufactured
  • Color Display
  • Three Year Warranty
  • Advanced algorithym for Faster, more Precise Measurements
  • RS-232/USB
  • Password Protection
  • GLP Compliant
The ProLab 3000 precision multi parameter meter enables you to perform pH, ORP, and ion-selective measurements rapidly and reliably with the ability to measure two parameters simultaneously plus temperature. This instrument is engineered/manufactured in Germany and backed by 65+ years of measuring experience. It provides the maximum degree of ease of use, reliability and, above all, measuring certainty for all applications. Fast, convenient and precise, the instrument can be operated just like a PC via the menu keys or by using a mouse (included). Easy to read, high resolution, backlit, color display.
The ProLab 3000 offers automatic user identification with electronic ID transponder and password as well as wireless sensor recognition. The CalClock provides a unique combination of sensor evaluation and calibration timer control in one easy to read symbol. Perfectly suitable for all GLP protocol. Check out all of our ProLab models to find the one that best fits your lab.
Certifications: CE Certified, UL STD 61010-1 Certified, cETLus Certified.
Ordering information: Meter and Universal Power Supply only. Kit also available (97042-270). Kit includes ProLab 3000 measuring instrument, mouse, IoLine 3in1 pharmaceutical grade double junction pH electrode (97042-384), buffer solutions, electrode arm, universal power supply and cover.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty