Optima™ TC120 Immersion Thermostat, General Purpose, Grant Instruments
TC120EURO
89260-848EA 1339 USD
89260-848
Optima™ TC120 Immersion Thermostat, General Purpose, Grant Instruments
The Optima™ TC120 general purpose, precision immersion thermostat can be combined with plastic baths, premium stainless steel baths or refrigeration units or used on its own with a clamp
Settable range is -20 to 120 °C (operation below ambient requires accessory cooling). Stirred circulation within the bath ensures excellent temperature stability and uniformity.
Optional clamp allows attachment of thermostat on to virtually any vertical sided bath or vessel (please order separately).
Digital microprocessor controlled the TC120 features a pump and connectors for external circulation, an easy to use rotary dial and two function keys for quick temperature setting and menu navigation. Also a bright, easy to view four digit display, three programmable temperature presets, low liquid protection, user adjustable over-temperature cut-out, visual alarm, countdown timer and 2 point user calibration facility for optimum accuracy.
Certifications: CE approved.