Optima™ T100 Immersion Thermostat, General Purpose, Grant Instruments
Supplier: Grant
T100 EURO
89260-846EA 461.25 USD
89260-846
Optima™ T100 Immersion Thermostat, General Purpose, Grant Instruments
Circulating Thermostats Open Circulator Systems Thermostat Controllers
The Optima™ T100 general purpose, precision immersion thermostat can be combined with plastic baths, premium stainless steel baths or refrigeration units or used on its own with a clamp
Settable range is 0 to 100 °C (operation below ambient requires accessory cooling). Stirred circulation within the bath ensures excellent temperature stability and uniformity.
Optional clamp allows attachment of thermostat on to virtually any vertical sided bath or vessel (please order separately).
Digital microprocessor controlled the T100 features an easy to use rotary dial and two function keys for quick temperature setting and menu navigation. Also, a bright, easy to view four digit display, three programmable temperature presets, low liquid protection, fixed over-temperature cut-out, visual alarm, countdown timer and 2 point user calibration facility for optimum accuracy.
Certifications: CE approved.