Pyocyanin ≥98% (by HPLC)

Supplier: Enzo Life Sciences
Danger

Synonyms: Sanasin1-hydroxy-N-methylphenazine

ENZ-53001-C001
89166-058EA 107.48 USD
89166-058
Pyocyanin ≥98% (by HPLC)
Pyocyanin

ROS inducer.


  • Reconstitute the contents of the vial in 100µl solvent to yield a 10mM stock solution.


Cytotoxic pigment secreted by Pseudomonas aeruginosa, which frequently infects the lungs of immuno-suppressed patients, as well as those with cystic fibrosis. Pyocyanin toxicity results from its ability to undergo reduction by NAD(P)H and subsequent generation of superoxide and H₂O₂ directly in the lungs.


Soluble in DMSO or dimethyl formamide.


Synonym: Sanasin, 1-hydroxy-N-methylphenazine

Formula: C₁₃H₁₀N₂O
MW: 210.2 g/mol
Storage Temperature: Ultra-Low Temperature Freezer (–80°C)
CAS Number: 85-66-5
https://us.vwr.com/stibo/web/std.lang.all/16/26/45401626.jpg

Order Now

Specification Test Results

Purity ≥98% (HPLC)
Formulation Lyophilised

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR