Would you like to log in again?
Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.
HPV 16 Control Slide, Enzo Life Sciences
Supplier: Enzo Life Sciences
ENZ-31877
89165-682EA 52.27 USD InStock
89165-682
HPV 16 Control Slide, Enzo Life Sciences
Slides and Cover Glasses Chamber Slides
The slide contains two wells of fixed CaSki cells with HPV 16 genomic DNA and is used as a positive control for DNA in situ hybridization of HPV-16.
No need for deparaffinization or protease treatment.
Order Now
ORDER
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty