Anti-GLP1 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
HYB-147-06-1 HYB-147-06-02
95054-354EA 545 USD
95054-354 95054-356
Anti-GLP1 Mouse Monoclonal Antibody
Glucagon-like peptide 1(7-36)amide (GLP-1(7-36)amide) is the principal active form of GLP-1, the other being GLP-1(7-37). GLP-1 is a peptide hormone of the glucagon family, produced by the L cells of the intestinal mucosa from the same prohormone as glucagon. The active forms are potent stimulators of glucose-dependent insulin secretion. The sequence of GLP-1 is fully conserved in all mammalian species examined so far.

  • For ELISA, Flow Cytometry, Immunohistochemistry

Recognizes mammalian GLP-1 (7-36, amide).

Type: Primary
Antigen: GLP-1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 ?
Reactivity: Mammals
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR