Anti-F9 Mouse Monoclonal Antibody

HYB-133-01-02
95054-318EA 545 USD
95054-318
Anti-F9 Mouse Monoclonal Antibody
Coagulation factor IX is a 57kDa single chain polypeptide, with a normal concentration in human plasma of approximately 3 µg/ml. Factor IX deficiency and lack of coagulation activity is seen in patients suffering from the innate X-chromosome linked disorder, haemophilia B.

  • For ELISA, Flow Cytometry, Western Blot

Recognizes human factor IX. Detects a band of ~57kDa by Western blot.

Type: Primary
Antigen: FACTOR IX
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 ?
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR