Anti-OVA Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
HYB-099-09-02
95054-118EA 545 USD
95054-118
Anti-OVA Mouse Monoclonal Antibody
Ovalbumin constitutes the main protein found in egg white, making up 60-65% of the total protein. Native chicken ovalbumin is a globular monomer with a molecular mass of 45kDa, structurally related to both serpins and lipocalins.

  • For ELISA, Flow Cytometry

Recognizes chicken ovalbumin. Detects a band of ~45kDa by Western blot.

Type: Primary
Antigen: OVA
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR