Anti-C2 Mouse Monoclonal Antibody

HYB-050-05-02
95053-950EA 545 USD
95053-950
Anti-C2 Mouse Monoclonal Antibody
Complement component C2 is a component of the classical complement pathway with involvement in the activation of C3 and C5. C2 consists of a single polypeptide chain with a molecular mass of 102kDa.

  • For ELISA, Flow Cytometry, Western Blot

Recognizes human complement component C2. Detects a band of ~102kDa by Western blot.

Type: Primary
Antigen: C2
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR