Anti-CFP Mouse Monoclonal Antibody (Biotin) [clone: 10C5]

HYB-039-04B-01
95053-930EA 907 USD
95053-930
Anti-CFP Mouse Monoclonal Antibody (Biotin) [clone: 10C5]
Properdin in plasma is a mixture of cyclic dimers, trimers and tetramers. The molecular weight of the glycosylated monomer is 53kDa. Properdin is a regulator protein of the alternative complement pathway. It stabilizes the C3 convertase, and binds to C3b in the C3bBb complex, thereby inhibiting cleavage of C3b by factor I and increasing the affinity for factor B. Serum concentration is approximately 25 �g/mL.

  • For ELISA, Flow Cytometry

Recognizes human properdin. Detects a band of ~53kDa by Western blot.

Type: Primary
Antigen: CFP
Clonality: Monoclonal
Clone: 10C5
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR