Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-C3 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
HYB-030-08-02
95053-918
Anti-C3 Mouse Monoclonal Antibody
C3 is a key component of the complement system, since classical and alternative activation pathways merge at the C3 activation step when C3 is split into C3a and C3b. The molecular mass of C3 is 185kDa, and it consists of two chains (110kDa and 75kDa) held together by disulfide bonds.

For ELISA, Flow Cytometry, Western Blot

Recognizes human complement component C3. Detects a band of ~185kDa by Western blot.

Type: Primary
Antigen: C3
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now


 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty