Human IgE

Supplier: Enzo Life Sciences
BPD-DIA-HE1-1
95053-726EA 2802.22 USD
95053-726
Human IgE
Antibodies
Purified human IgE with kappa light chains produced in vitro from a monoclonal hybridoma.

  • Non-Immune
  • Native Protein
  • Used in Immunocytochemistry

Type:
Antigen:
Clonality:
Clone:
Conjugation:
Epitope:
Host:
Isotype: IgE
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR