Anti-GLP1 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
ABS-044-53-1
95053-544EA 3970.36 USD
95053-544
Anti-GLP1 Mouse Monoclonal Antibody
Antibodies
Glucagon-like peptide-1 (9-36) amide and GLP-1 (9-37) are the forms of GLP-1 which result from rapid degradation of the active forms of the peptide (GLP-1 (7-36) amide and GLP-1 (7-37)) by the enzyme dipeptidyl peptidase-IV (DPP-IV, also known as CD26 or adenosine deaminase binding protein). GLP-1 is a peptide hormone of the glucagon family, produced by the L cells of the intestinal mucosa from the same prohormone as glucagon. The active forms are potent stimulators of glucose-dependent insulin secretion. The sequence of GLP-1 is fully conserved in all mammalian species examined so far.

  • For ELISA, Flow Cytometry, Western Blot

Recognizes mammalian GLP-1 (9-37, 9-36, amide, free NT-specific).

Type: Primary
Antigen: GLP-1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 ?
Reactivity: Mammals
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR