Anti-Aprotinin Mouse Monoclonal Antibody (Biotin)

Supplier: Enzo Life Sciences
ABS 037-18B005
CA95056-586
Anti-Aprotinin Mouse Monoclonal Antibody (Biotin)
Antibodies
Aprotinin, also called bovine pancreatic trypsin inhibitor, is a single chain globular Kunitz-type serine proteinase inhibitor found in bovine pancreas, parotid gland and lung. It forms stable, enzymatically inactive complexes with trypsin, chymotrypsin, plasmin and kallikrein. Aprotinin has been used therapeutically as an antifibrinolytic agent under the name Trasylol (Bayer) to reduce blood loss during cardiopulmonary bypass surgery, but this use has been associated with an increased frequency of acute renal injury.

  • For ELISA, Flow Cytometry

Recognizes bovine aprotinin.

Type: Primary
Antigen: [delete]
Clonality: Monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR