Anti-ADM Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
ABS-027-01-1 ABS-027-01-02
95053-358EA 3894.79 USD
95053-358 95053-356
Anti-ADM Mouse Monoclonal Antibody
Antibodies
Human adrenomedullin is a 52 amino-acid peptide (mw 6029) with the physiological effect of lowering blood pressure through vasodilatation, an effect it shares with calcitonin gene-related peptide (CGRP) and vasoactive intestinal peptide (VIP). In addition, adrenomedullin produces cardiovascular effects such as growth and development of cardiovascular tissues and bone, and effects in the central nervous system connected with the regulation of fluid balance.

  • For ELISA, Flow Cytometry, Western Blot

Recognizes human adrenomedulin. Detects a band of ~6kDa by Western blot.

Type: Primary
Antigen: ADM
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR