Anti-Exendin-4 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
ABS-012-24-1
95053-344EA 3970.36 USD
95053-344
Anti-Exendin-4 Mouse Monoclonal Antibody
Antibodies
Exendin-4 is a 39-amino-acid peptide found in venom from the Gila monster Heloderma suspectum. It is a member of the glucagon-secretin family of peptide hormones and neuropeptides. Exendin-4 is a potent agonist of the GLP-1 receptor and hence a potent stimulator of insulin secretion.

  • For ELISA, Flow Cytometry

Recognizes human exendin-4. Detects a band of ~4kDa by Western blot.

Type: Primary
Antigen: EXENDIN-4
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 ?
Reactivity: Heloderma Suspectum
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR