Z-Arg-Leu-Arg-Gly-Gly-AMC
Supplier: Enzo Life Sciences
ZW8585-0005
89165-656EA 230.88 USD
89165-656
Z-Arg-Leu-Arg-Gly-Gly-AMC
Proteins and Peptides
A peptide-AMC substrate for isopeptidase T based on the C-terminal residues of ubiquitin.