N-Arachidonoyldopamine ≥98% (by TLC)
Supplier: Enzo Life Sciences
BML-VR104-0025
89165-502EA
296.03
USD
89165-502
N-Arachidonoyldopamine ≥98% (by TLC)
N-Arachidonoyldopamine
Activator of CB1 receptor and TRPV1
Endogenous, specific ligand for the CB1 receptor (CB1: Ki=250nM; CB2: Ki=12µM) [1] and TRPV1 [2] (EC50=50nM) found in nervous tissues [3, 4]. Immunosuppressant inhibiting T cell proliferation and phosphorylation of the NF-κB p65 subunit [5]. Potent vasorelaxant [6] and inhibitor of HIV-1 [7].
Formula:
C₂₈H₄₁NO₃ MW: 439.64 g/mol |
MDL Number:
MFCD03412031 CAS Number: 199875-69-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...