Anti-VINC Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
VG6110-0100
89165-482EA 265.01 USD
89165-482
Anti-VINC Mouse Monoclonal Antibody
Antibodies
Vinculin is associated with the cytoplasmic aspect of adhesion points and acts as a link between the ends of actin microfilaments and the plasma membrane. Vinculin may also be involved in the transmembrane induction of actin-bundle formation.

  • For Immunohistochemistry, Western Blot

Recognizes human Vinculin.

Type: Primary
Antigen: VINC
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR