Anti-Vasoactive Intestinal (Poly)Peptide Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
VA1285-0025 VA1285-0100
89165-474EA 131.6 USD
89165-474 89165-476
Anti-Vasoactive Intestinal (Poly)Peptide Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit

  • For Immunohistochemistry

Recognizes human, rat and pig VIP.

Type: Primary
Antigen: VASOACTIVE INTESTINAL (POLY)PEPTIDE
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR