Human Recombinant UbcH13/Mms2 (from E. coli), HIS Tag
Supplier: Enzo Life Sciences
UW9565-0100
89165-392EA 380.59 USD
89165-392
Human Recombinant UbcH13/Mms2 (from E. coli), HIS Tag
Proteins and Peptides
Produced in E. coli.
A heterodimer composed of the catalytically active ubiquitin-conjugating enzyme UbcH13 and its catalytically inactive paralog hMms2, which lacks an active site cysteine residue.