Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

UbcH5b (human), (recombinant) (His-tag), Enzo Life Sciences

Supplier: Enzo Life Sciences
UW9060-0100 UW9060-1000
Produced in E. coli.

A number of E2s in Saccharomyces cerevisiae and their homologues have been identified. One such family of E2s is the UBC4/5, characterised as essential for the degradation of short-lived, regulatory and abnormal proteins. Protein levels of S. cerevisiae UbC4/5 are up-regulated in response to stress, and their loss results in severe effects on cellular functions. A human gene product that is 79% identical to S. cerevisiae UBC4/5 in amino-acid sequence was identified as UbcH5a. In addition, two other human members of this highly conserved E2 class were also cloned and designated as UbcH5b and UbcH5c, having 88% and 89% identity to UbcH5a, respectively. Members of the UbcH5a/b/c are the most active class of E2s in cell extracts. The importance of this enzyme class is underscored by the critical role of UBC4/5 in S. cerevisiae. UbcH5a stimulates the conjugation of ubiquitin to the tumour-repressor p53 in the presence of E6-AP and E6. Moreover, UbcH5 family is implicated in c-fos recognition, the modulation of which is controlled by the ubiquitin-proteasome pathway. UbcH5b and UbcH5c are associated with the signal-induced conjugation and subsequent degradation of IkBalpha in the presence of the SCF complexes. UbcH5c also catalyses the ubiquitination leading to the processing of p105 precursor to form p50, a subunit of the heterodimeric transcription factor NF-kB. The range and diversity of substrates and E3s with which this class of E2 enzymes interact, suggest their complex roles in cellular functions require to be studied further.




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µg UW9060-0100 89165-318 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

1 mg UW9060-1000 89165-320 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Protein/Peptide Name UBCH5B (His-tag)
Protein/Peptide Type Recombinant
Species Human
Source E. coli
Purity ≥95% (SDS-PAGE)
Formulation Liquid. In TRIS-HCl, pH7.5, containing 0.5mM DTT

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty