Ubiquitin
Supplier: Enzo Life Sciences
UW8630-0500
89165-270EA 279.88 USD
89165-270
Ubiquitin
Proteins and Peptides
Ubiquitin is coupled to agarose, via the epsilon amino groups of its internal lysine residues, leaving the C-terminus free. The loading is approximately10mg protein/ml of settled resin.