Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

POLYSUMO-QAPTURE® Kit, Enzo Life Sciences

Supplier: Enzo Life Sciences
UW0955-0001
This Kit is an efficient tool for the selective isolation of polySUMOylated proteins. The kit facilitates the affinity purification of polySUMOylated proteins from cell extracts and tissue lysates using a high-binding multi-SIM-containing affinity matrix, whilst discriminating against free and mono-SUMO-conjugate binding.

  • Fast, simple, and reliable assay
  • High specificity, high through-put capacity
  • Amenable to analysis via Western blotting or proteomic methods

For the isolation and enrichment of polySUMOylated proteins

Captured proteins are specifically eluted prior to analysis by Western blotting, using the SUMO2/3 antibody provided or antibodies to specific proteins of interest, further purification by substrate specific immunoprecipitation, biochemical characterization, or potential substrate identification by proteomic methods polySUMO modified proteins by proteomic analysis following release of free polySUMOylated proteins in their active/native form by specific elution from the affinity matrix. Selective purification/pull down of polySUMOylated proteins from in vitro SUMOylation assays.

PolySUMO-conjugate-containing samples are prepared in native form in the presence of protease inhibitors to prevent loss through the action of deSUMOulating enzymes. Optimisation of binding permits complete isolation of the full range of polySUMO-protein conjugates from a specific lysate.

The kit is also supplied with a high quality polySUMOylated protein solution for use as a positive control in POLYSUMO-QAPTURE® assays.

Capture and isolation of polySUMOylated protein conjugates from cell/tissue lysates of interest with subsequent detection and analysis by Western blotting.

Ordering information: Quantity: Sufficient for 10 binding assays.
Contents: PolySUMO-Qapture matrix (Prod. No. BML-UW0995) 200µL settled resin provided, 50% suspension.
Control SUMOylated-protein lysate (Prod. No. BML-UW0985) 220 µg (110 µL), 2mg/mL in 50mM TRIS-Cl, pH 7.5, 150mM sodium chloride, 1% NP-40, 0.5% deoxycholate, 200mM iodoacetamide.
PolySUMO-Qapture elution buffer (Prod. No. BML-UW1005) 1mL.
SUMO-2/3, sheep polyclonal antibody (Prod. No. BML-KW0510) 10µL.
0.5mL Screw capped tube pack (Prod. No. BML-UW0990) 10 Tubes.




 

ORDER

Expand All / Collapse All
Size Sufficient For Supplier No. VWR Catalog Number Unit Price Quantity
1 Kit 10 binding assays UW0955-0001 89165-246 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty