Nona-Ubiquitin (linear) (from E. coli)
Supplier: Enzo Life Sciences
UW0810-0100
89165-224EA 186.22 USD
89165-224
Nona-Ubiquitin (linear) (from E. coli)
Proteins and Peptides
Produced in E. coli.
Linear polyubiquitin chains are useful tools for investigating, amongst other things, the specificity and reactivity of deubiquitinylating enzymes (DUBs) and the recognition and interaction of linear polyubiquitin modified proteins with by ubiquitin binding domain (UBDs) containing proteins.