Hepta-Ubiquitin (linear) (from E. coli)
Supplier: Enzo Life Sciences
UW0800-0100
89165-220EA 186.22 USD
89165-220
Hepta-Ubiquitin (linear) (from E. coli)
Proteins and Peptides
Linear polyubiquitin chains are useful tools for investigating, amongst other things, the specificity and reactivity of deubiquitinylating enzymes (DUBs) and the recognition and interaction of linear polyubiquitin modified proteins with by ubiquitin binding domain (UBDs) containing proteins.