([K33-only]Ub)n-ubiquitinylated substrate, Enzo Life Sciences
UW0635-0025
89165-194EA 276.19 USD
89165-194
([K33-only]Ub)n-ubiquitinylated substrate, Enzo Life Sciences
Protein components produced in E. coli. Synthesized enzymatically in vitro. (K33-only)Ubn-ubiquitinylated substrate protein was produced using K33-only mutant ubiquitin.