Human Recombinant SUMO3, Agarose
Supplier: Enzo Life Sciences
UW0105-0500
89165-098EA 228.73 USD
89165-098
Human Recombinant SUMO3, Agarose
Human recombinant SUMO-3 coupled to agarose at 0.5mg protein per ml settled resin.
For isolation and enrichment of SUMO-3 interacting proteins. SUMO-3 agarose may have considerable utility for the capture/pull down of SUMO-3 interacting partners, including processing/signalling pathway proteins and SUMO-3 modification substrates, from cell lysates and extracts by affinity binding. The immobilized protein retains the exposed C-terminal diglycine required for SUMO-3 modification and therefore may be suitable for in vitro SUMOylation. Captured proteins can be eluted readily for identification, analysis or subsequent biochemical reaction.