Ansamitocin P-3 ≥93% (by HPLC)
Supplier: Enzo Life Sciences
BML-T121-0001
89165-000EA
485.46
USD
89165-000
Ansamitocin P-3 ≥93% (by HPLC)
Ansamitocin P-3
Ansamitocin P-3 is a maytansine analog which displays potent cytotoxicity against the human solid tumor cell lines A-549 and HT-29. It shares the same binding site as rhizoxin and phomopsin A on porcine brain tubulin. Maytansine and its analogs cause extensive disassembly of the microtubule and totally prevent tubulin spiralization.
- Microtubule inhibitor
White to off-white crystalline solid.
Soluble in DMSO.
Formula:
C₃₂H₄₃ClN₂O₉ MW: 635.1 g/mol Storage Temperature: Freezer |
CAS Number:
66547-09-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...