Suc-Arg-Pro-Phe-His-Leu-Leu-Val-Tyr-AMC
Supplier: Enzo Life Sciences
SW8525-0005
89164-940EA 449.24 USD
89164-940
Suc-Arg-Pro-Phe-His-Leu-Leu-Val-Tyr-AMC
Proteins and Peptides
A fluorogenic substrate for renin and proteinase A, also a substrate for the chymotrypsin-like activity of the 20S proteasome.