BSA (dimethylated), Enzo Life Sciences
BML-SW126-0100
89164-938EA 76 USD
89164-938
BSA (dimethylated), Enzo Life Sciences
Methylated bovine serum albumin for use as a Western positive control or for peptide blocking experiments for antibodies to dimethyl lysine (Prod. No. BML-SA667 and BML-SA668).