Jurkat Cell Extract (Induced), Enzo Life Sciences

Supplier: Enzo Life Sciences
BML-SW116-0100
89164-926EA 54.25 USD
89164-926
Jurkat Cell Extract (Induced), Enzo Life Sciences
Cells
Immunoblotting standard

  • Formulation: Liquid
  • Storage Conditions: –80 °C (Long Term)

Applications include Western Blotting control for PARP antibodies to detect intact 116 kDA PARP and 85 kDa apoptosis-related cleavage product and detection of procaspase-3 processing to active enzyme using caspase-3 antibody and pro-caspase-9.

Whole cell extract of human Jurkat macrophage cells, induced to undergo apoptosis by the chemotherapeutic agent etoposide.

Delivery information: Supplied with SDS-PAGE Buffer.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR