Rat Brain Tissue Extract, Enzo LifeScience
Supplier: Enzo Life Sciences
BML-SW103-0200
89164-906EA 45.02 USD
89164-906
Rat Brain Tissue Extract, Enzo LifeScience
Specimens Preserved Specimens Preserved Animal Materials
Whole brain protein extract from rat
Supplied as tissue extract (approximately 5 mg/mL) in SDS-PAGE sample buffer, ready-to-use at 5 µL/lane. Useful as a Western blotting control for Anti-nNOS and PKA RIIß antibody, or for use with other antibodies.